missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,4-dichlorobenzene, 98+%

Product Code. 10543221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10543221 100 g Glass bottle 100g
10499940 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Catalog No. 10543221 Lieferant Thermo Scientific Acros Lieferanten-Nr. 269811000
 Restricted Item
Estimated Shipment: 07-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 18282-59-2 | C6H3BrCl2 | 225.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 18282-59-2
Molecular Formula C6H3BrCl2
Molecular Weight (g/mol) 225.89
MDL Number MFCD00040849
InChI Key CFPZDVAZISWERM-UHFFFAOYSA-N
Synonym 1-bromo-3,4-dichlorobenzene, 3,4-dichlorobromobenzene, benzene, 4-bromo-1,2-dichloro, 3,4-dichlorophenyl bromide, 3,4-dichloro-1-bromobenzene, 4-bromo-1,2-dichloro-benzene, 5-bromdichlorbenzol, pubchem3611, acmc-1btol, 3,4-dichloro-bromobenzene
PubChem CID 29013
IUPAC Name 4-bromo-1,2-dichlorobenzene
SMILES ClC1=CC=C(Br)C=C1Cl

Spezifikation

CAS Min % 98.0
CAS Max % 100.0
Melting Point 23.0°C to 24.0°C
Color Yellow
Boiling Point 140.0°C to 142.0°C (30.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula BrC6H3Cl2
Refractive Index 1.5986 to 1.6006
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 225.91
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 1-Bromo-3, 4-dichlorobenzene, 98+%
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • 1-Bromo-3,4-dichlorobenzene
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Skin sensitiser Category 1
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.