Learn More
1-Bromo-3,5-bis(trifluoromethyl)benzene, 98%
Descrizione
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Bis(trifluoromethyl)-5-bromobenzene is used to prepare the tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ion, which acts as a stabilizing counterion for electrophilic organic and organometallic cations.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifica
Specifica
| Melting Point | -16°C |
| Density | 1.698 |
| Boiling Point | 152°C to 155°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.427 |
| Quantity | 250 g |
| Beilstein | 2123669 |
| Solubility Information | Immiscible with water. |
| Formula Weight | 293.01 |
| Percent Purity | 98% |
| Vedi altri risultati |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.