missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloro-2-fluorobenzene, 95%

Product Code. 15498168
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498168 100 g 100g
15488168 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498168 Supplier Thermo Scientific Alfa Aesar Supplier No. H61622.22
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 144584-65-6 | C6H3BrClF | 209.442 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144584-65-6
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD03412182
InChI Key DRNWNTAANHEQMK-UHFFFAOYSA-N
Synonym 3-chloro-2-fluorobromobenzene, 1-bromo-2-fluoro-3-chlorobenzene, 1-bromo-3-chloro-2-fluoro-benzene, pubchem4338, acmc-1c0jw, intermediates-zcf02109, ksc493i6p, benzene,1-bromo-3-chloro-2-fluoro
PubChem CID 3685762
IUPAC Name 1-bromo-3-chloro-2-fluorobenzene
SMILES C1=CC(=C(C(=C1)Br)F)Cl

Specifications

Melting Point 42°C to 46°C
Color White to Yellow
Quantity 100 g
Formula Weight 209.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1-Bromo-3-chloro-2-fluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.