missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-chloro-5-fluorobenzene, 98%, Thermo Scientific™

Product Code. 13418618 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13418618 25 g 25g
13428618 5 g 5g
13438618 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13418618 Supplier Thermo Scientific Acros Supplier No. 448750250
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 33863-76-2
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.45
InChI Key GGMDFPMASIXEIR-UHFFFAOYSA-N
Synonym 3-chloro-5-fluorobromobenzene, 1-bromo3-chloro-5-fluorobenzene, 1-bromo-3-chloro-5-fluoro-benzene, benzene, 1-bromo-3-chloro-5-fluoro, 1,3,5-fluorochlorobromobenzene, pubchem2261, acmc-209i3i, ksc494q7t, 3-chloro-5-fluoro-bromobenzene, 1-bromo-3-chloro-5-flurobenzene
PubChem CID 2736223
IUPAC Name 1-bromo-3-chloro-5-fluorobenzene
SMILES C1=C(C=C(C=C1Cl)Br)F

Specifications

Density 1.7200g/mL
Boiling Point 72°C (5.0 mmHg)
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(Cl)F
Quantity 25 g
Specific Gravity 1.72
Formula Weight 209.45
Percent Purity 98%
Chemical Name or Material 1-Bromo-3-chloro-5-fluorobenzene
compliance-icons
Product Identifier
  • 1-Bromo-3-chloro-5-fluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.