missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-cyclopropylbenzene, 96%, Thermo Scientific™

Product Code. 15239806 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15239806 250 mg 250mg
15229806 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15239806 Supplier Thermo Scientific Acros Supplier No. 452582500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1798-85-2
Molecular Formula C9H9Br
Molecular Weight (g/mol) 197.08
MDL Number MFCD01070823
InChI Key BCBNVQYWODLZSI-UHFFFAOYSA-N
Synonym m-bromophenylcyclopropane, 3-bromo-1-cyclopropylbenzene, 1-bromo-3-cyclopropyl-benzene, benzene, 1-bromo-3-cyclopropyl, 3-cyclopropylbromobenzene, pubchem22120, bromo-3-cyclopropyl-benzene, 1-cyclopropyl-3-bromobenzene, 1-bromo-3-cyclopropyl benzene, benzene,1-bromo-3-cyclopropyl
PubChem CID 3282672
IUPAC Name 1-bromo-3-cyclopropylbenzene
SMILES BrC1=CC=CC(=C1)C1CC1

Specifications

Assay Percent Range 95% min. (GC)
Quantity 250 mg
Formula Weight 197.07
Percent Purity 96%
Chemical Name or Material 1-Bromo-3-cyclopropylbenzene
compliance-icons
Product Identifier
  • 1-Bromo-3-cyclopropylbenzene, 96%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.