missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-ethylbenzene, 97%, Thermo Scientific™

Product Code. 10150892
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10150892 1 g 1g
10283372 10 g 10g
10542083 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10150892 Supplier Thermo Scientific Maybridge Supplier No. TL00571DA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2725-82-8
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00156128
InChI Key ZRFJYAZQMFCUIX-UHFFFAOYSA-N
Synonym benzene, 1-bromo-3-ethyl, 3-ethylbromobenzene, 1-bromo-3-ethyl-benzene, 3-bromoethyl benzene, 3ethylbromobenzene, m-bromoethylbenzene, 3-bromo-ethylbenzene, pubchem3771, 1bromo-3-ethylbenzene, 1-bromo-3-ethylbenze
PubChem CID 123170
IUPAC Name 1-bromo-3-ethylbenzene
SMILES CCC1=CC(=CC=C1)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula CH3CH2C6H4Br
Quantity 1 g
Formula Weight 185.06
Percent Purity 97%
Chemical Name or Material 1-Bromo-3-ethylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.