missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-methyl-2-butene, 90+%, stab. with silver

Product Code. 11435383
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435383 10 g 10g
11445383 50 g 50g
11455383 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11435383 Supplier Thermo Scientific Alfa Aesar Supplier No. A13388.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 870-63-3 | C5H9Br | 149.031 g/mol

1-Bromo-3-methyl-2-butene is considered as a source of 3,3-dimethylallyl group and used in the preparation of natural products. It is also employed in the synthesis of base-modified pyrimidine nucleosides. Further, it is used in the preparation of (±)-eldanolide, 1-(3,3-dimethylallyl)-L-tryptophan and (±)-fumagillin, an antibiotic isolated from Aspergillus fumigattus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-3-methyl-2-butene is considered as a source of 3,3-dimethylallyl group and used in the preparation of natural products. It is also employed in the synthesis of base-modified pyrimidine nucleosides. Further, it is used in the preparation of (±)-eldanolide, 1-(3,3-dimethylallyl)-L-tryptophan and (±)-fumagillin, an antibiotic isolated from Aspergillus fumigattus.

Solubility
Immiscible with water

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-63-3
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.031
MDL Number MFCD00000242
InChI Key LOYZVRIHVZEDMW-UHFFFAOYSA-N
Synonym 1-bromo-3-methyl-2-butene, 3,3-dimethylallyl bromide, prenyl bromide, 3-methyl-2-butenyl bromide, 2-butene, 1-bromo-3-methyl, dimethylallyl bromide, 1-bromo-3-methyl-but-2-ene, .gamma.,.gamma.-dimethylallyl bromide, 3-methylcrotyl bromide, 3,3-dimethylallylbromide
PubChem CID 70092
IUPAC Name 1-bromo-3-methylbut-2-ene
SMILES CC(=CCBr)C

Specifications

Density 1.281
Boiling Point 135°C
Flash Point 32°C (90°F)
Odor Pungent
Refractive Index 1.4895
Quantity 10 g
UN Number UN2920
Beilstein 1420778
Sensitivity Light sensitive
Solubility Information Immiscible with water
Formula Weight 149.03
Percent Purity ≥90%
Chemical Name or Material 1-Bromo-3-methyl-2-butene, stabilized with silver
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.