missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-methyl-2-butene, 96%

Product Code. 15254704 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15254704 50 g 50g
10105850 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15254704 Supplier Thermo Scientific Acros Supplier No. 307410500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 870-63-3 | C5H9Br | 149.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-63-3
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.04
MDL Number MFCD00000242
InChI Key LOYZVRIHVZEDMW-UHFFFAOYSA-N
Synonym 1-bromo-3-methyl-2-butene, 3,3-dimethylallyl bromide, prenyl bromide, 3-methyl-2-butenyl bromide, 2-butene, 1-bromo-3-methyl, dimethylallyl bromide, 1-bromo-3-methyl-but-2-ene, .gamma.,.gamma.-dimethylallyl bromide, 3-methylcrotyl bromide, 3,3-dimethylallylbromide
PubChem CID 70092
IUPAC Name 1-bromo-3-methylbut-2-ene
SMILES CC(=CCBr)C

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Brown or Yellow
Density 1.2700g/mL
Boiling Point 82.0°C to 83.0°C (150.0 mmHg)
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (ex Br) (Argentometry)
Linear Formula BrCH2CH=C(CH3)2
Refractive Index 1.4890 to 1.4910
Quantity 50 g
UN Number 2924
Specific Gravity 1.27
Stabilizer silver wool
Formula Weight 149.04
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Bromo-3-methyl-2-butene, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-3-methyl-2-butene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1-Bromo-3-methyl-2-butene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.