missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-nitrobenzene, 99%

Codice prodotto. 10571601 Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Cambia vista
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Dimensione della confezione:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Packaging Dimensione della confezione
10571601 25 g Plastic bottle 25g
10163020 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10571601 Fornitore Thermo Scientific Acros N. del fornitore 160390250
 Restricted Item
Estimated Shipment: 19-01-2027
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 585-79-5 | C6H4BrNO2 | 202.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 585-79-5
Molecular Formula C6H4BrNO2
Molecular Weight (g/mol) 202.01
MDL Number MFCD00024298
InChI Key FWIROFMBWVMWLB-UHFFFAOYSA-N
Synonym 3-bromonitrobenzene, m-bromonitrobenzene, benzene, 1-bromo-3-nitro, 3-nitrobromobenzene, m-nitrobromobenzene, 1-bromo-3-nitro-benzene, 3-bromo-1-nitrobenzene, ccris 3114, meta-bromonitrobenzene, bromonitrobenzene-3
PubChem CID 11457
IUPAC Name 1-bromo-3-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(Br)=C1

Specifica

CAS Min % 98.5
CAS Max % 100.0
Melting Point 51°C to 54°C
Color Yellow to Brown
Density 1.7040g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula BrC6H4NO2
Quantity 25 g
Beilstein 05, 248
Formula Weight 202.01
Percent Purity ≥98.5%
Physical Form Crystalline Solid
Chemical Name or Material 1-Bromo-3-nitrobenzene, 99%
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • 1-Bromo-3-nitrobenzene
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.