missing translation for 'onlineSavingsMsg'
Learn More

1-bromo-3-phenoxybenzene, Thermo Scientific™

Product Code. 11336136
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11336136 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11336136 Supplier Thermo Scientific Maybridge Supplier No. MAY00156DA
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 6876-00-2
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Molecular Formula C12H9BrO
MDL Number MFCD00474520
Quantity 1 g
Synonym 3-phenoxybromobenzene, 3-bromodiphenyl ether, benzene, 1-bromo-3-phenoxy, benzene,1-bromo-3-phenoxy, m-bromodiphenyl ether, 3-phenoxyphenyl bromide, 1-phenoxy-3-bromobenzene, 3-bromo-1-phenoxybenzene, acmc-209o4k, 1-bromo-3-phenoxy-benzene
InChI Key AHDAKFFMKLQPTD-UHFFFAOYSA-N
SMILES BrC1=CC=CC(OC2=CC=CC=C2)=C1
IUPAC Name 1-bromo-3-phenoxybenzene
Molecular Weight (g/mol) 249.11
PubChem CID 96165
Formula Weight 249.11
Percent Purity 97%
Chemical Name or Material 1-Bromo-3-phenoxybenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.