missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-butoxybenzene, 97%, Thermo Scientific™

Product Code. 10661663
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10661663 50 g 50g
10096804 5 g 5g
10140722 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10661663 Supplier Thermo Scientific Maybridge Supplier No. BTB09793EE
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 39969-57-8
Molecular Formula C10H13BrO
Molecular Weight (g/mol) 229.117
MDL Number MFCD00037100
InChI Key BOUVKHWPQNEXTO-UHFFFAOYSA-N
PubChem CID 142395
IUPAC Name 1-bromo-4-butoxybenzene
SMILES CCCCOC1=CC=C(C=C1)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula BrC6H4O(CH2)3CH3
Quantity 50 g
Synonym p-bromophenyl butyl ether, benzene,1-bromo-4-butoxy, p-butoxybromobenzene, benzene, 1-bromo-4-butoxy, 4-bromo-1-butoxybenzene, 4-butoxybromobenzene, 4-n-butoxyphenyl bromide, 1-butoxy-4-bromobenzene, 4-bromophenyl butyl ether
Formula Weight 229.12
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-butoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.