missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-chlorobutane, 99%

Product Code. 11430233
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430233 25 g 25g
11440233 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430233 Supplier Thermo Scientific Alfa Aesar Supplier No. A11399.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6940-78-9 | C4H8BrCl | 171.462 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6940-78-9
Molecular Formula C4H8BrCl
Molecular Weight (g/mol) 171.462
MDL Number MFCD00001010
InChI Key NIDSRGCVYOEDFW-UHFFFAOYSA-N
Synonym butane, 1-bromo-4-chloro, tetramethylene chlorobromide, 4-bromo-1-chlorobutane, 1-chloro-4-bromobutane, 4-bromobutyl chloride, 4-chlorobutyl bromide, 1-brom-4-chlorbutan, 4-chlorobromobutane, bromo 4 chlorobutane, bromo-4-chlorobutane
PubChem CID 81364
IUPAC Name 1-bromo-4-chlorobutane
SMILES C(CCBr)CCl

Specifications

Density 1.489
Boiling Point 175°C to 176°C
Flash Point 60°C (140°F)
Odor Aromatic
Refractive Index 1.4875
Quantity 25 g
UN Number UN1993
Beilstein 1098275
Formula Weight 171.47
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-chlorobutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.