missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-cyclohexylbenzene, 98%

Product Code. 11301647
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11301647

Brand: Thermo Scientific Alfa Aesar L04636.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25109-28-8 | C12H15Br | 239.156 g/mol

1-Bromo-4-cyclohexylbenzene is used as a organic light-emitting diode and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-cyclohexylbenzene is used as a organic light-emitting diode and as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25109-28-8
Molecular Formula C12H15Br
Molecular Weight (g/mol) 239.156
MDL Number MFCD00019339
InChI Key LVIJLEREXMVRAN-UHFFFAOYSA-N
Synonym p-bromocyclohexylbenzene, 1-brom-4-cyclohexylbenzol, zlchem 88, 4-cyclohexylbromobenzene, 4-bromophenylcyclohexane, acmc-1ccsm, 4-bromo-1-cyclohexylbenzene, 1-bromo-4-cyclohexyl-benzene, benzene, 1-bromo-4-cyclohexyl, lvijlerexmvran-uhfffaoysa
PubChem CID 90718
IUPAC Name 1-bromo-4-cyclohexylbenzene
SMILES C1CCC(CC1)C2=CC=C(C=C2)Br

Specifications

Density 1.287
Boiling Point 90°C to 94°C (0.01 mmHg)
Refractive Index 1.5595
Quantity 25 g
Beilstein 2209869
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 239.16
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-cyclohexylbenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.