missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-ethylbenzene, 99%

Product Code. 11429923
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429923 50 g 50g
11419923 10 g 10g
11439923 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429923 Supplier Thermo Scientific Alfa Aesar Supplier No. A13013.18
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1585-07-5 | C8H9Br | 185.064 g/mol

1-Bromo-4-ethylbenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-4-ethylbenzene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Avoid excessive heat and light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1585-07-5
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000111
InChI Key URFPRAHGGBYNPW-UHFFFAOYSA-N
Synonym p-bromoethylbenzene, benzene, 1-bromo-4-ethyl, 4-ethylbromobenzene, p-ethylbromobenzene, 1-bromo-4-ethyl-benzene, unii-42li8tat6j, 1-ethyl-4-bromobenzene, 4-bromo-1-ethylbenzene, 4-ethyl-1-bromobenzene, 42li8tat6j
PubChem CID 15313
IUPAC Name 1-bromo-4-ethylbenzene
SMILES CCC1=CC=C(C=C1)Br

Specifications

Density 1.341
Boiling Point 203°C to 204°C
Flash Point 63°C (145°F)
Refractive Index 1.544
Quantity 50 g
Beilstein 1856989
Solubility Information Insoluble in water.
Formula Weight 185.06
Percent Purity 99%
Chemical Name or Material 1-Bromo-4-ethylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.