missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(ethylthio)benzene, 97%

Product Code. 11343207
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343207 100 g 100g
11323207 5 g 5g
11333207 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11343207 Supplier Thermo Scientific Alfa Aesar Supplier No. L13718.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 30506-30-0 | C8H9BrS | 217.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30506-30-0
Molecular Formula C8H9BrS
Molecular Weight (g/mol) 217.124
MDL Number MFCD01318109
InChI Key CTFHVAKXUMMGOC-UHFFFAOYSA-N
Synonym 1-bromo-4-ethylthio benzene, 1-bromo-4-ethylsulfanyl benzene, 4-bromophenyl ethyl sulfane, benzene,1-bromo-4-ethylthio, acmc-1ae1n, 1-bromo-4-ethylthiobenzene, 4-bromo-1-ethylthiobenzene, ethyl 4-bromophenyl sulfide, 1-bromo-4-ethylthio-benzene, 1-bromo-4-ethylsulfanyl-benzene
PubChem CID 7006650
IUPAC Name 1-bromo-4-ethylsulfanylbenzene
SMILES CCSC1=CC=C(C=C1)Br

Specifications

Boiling Point 109°C to 111°C (2 mmHg)
Refractive Index 1.6069
Quantity 100 g
Beilstein 2354970
Formula Weight 217.13
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-(ethylthio)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.