missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-ethynylbenzene, 98%

Product Code. 11364272 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11364272 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11364272 Supplier Thermo Scientific Acros Supplier No. 442790010
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 766-96-1 | C8H5Br | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 766-96-1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 64°C to 65°C
Color White to Yellow
Boiling Point 71°C to 72°C (3.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H5Br
Quantity 1 g
Synonym 4-bromophenylacetylene, benzene, 1-bromo-4-ethynyl, 1-bromo-4-ethynyl-benzene, 4-bromophenyl acetylene, 4'-bromophenyl acetylene, 4'-bromophenylacetylene, p-bromophenylacetylene, p-bromophenyl acetylene, 4-bromphenyl acetylene, 4-bromo-1-ethynylbenzene
InChI Key LTLVZQZDXQWLHU-UHFFFAOYSA-N
SMILES C#CC1=CC=C(C=C1)Br
IUPAC Name 1-bromo-4-ethynylbenzene
Molecular Weight (g/mol) 181.03
PubChem CID 136603
Formula Weight 181.03
Percent Purity 98%
Physical Form Crystalline Powder, Powder, or Solid
Chemical Name or Material 1-Bromo-4-ethynylbenzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-4-ethynylbenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.