missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-iodonaphthalene, 98%

Product Code. 15482199
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15482199 1 g 1g
15402209 250 mg 250mg
15492199 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15482199 Supplier Thermo Scientific Alfa Aesar Supplier No. H64825.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 63279-58-3 | C10H6BrI | 332.966 g/mol

It is used as an OLED material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an OLED material.

Solubility
Slightly soluble in water. (3.6 g/L) (25°C)

Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63279-58-3
Molecular Formula C10H6BrI
Molecular Weight (g/mol) 332.966
MDL Number MFCD01166242
InChI Key HQHHKYXPFKHLBF-UHFFFAOYSA-N
Synonym 1-iodo-4-bromonaphthalene, 1-iodo-4-bromoaphthalene, 1-bromo-4-iodonaphth, naphthalene, 1-bromo-4-iodo
PubChem CID 5073667
IUPAC Name 1-bromo-4-iodonaphthalene
SMILES C1=CC=C2C(=C1)C(=CC=C2I)Br

Specifications

Melting Point 84°C to 89°C
Quantity 1 g
Beilstein 2044950
Sensitivity Light sensitive
Solubility Information Slightly soluble in water. (3.6g/L) (25°C)
Formula Weight 332.97
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Bromo-4-iodonaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.