missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)-3-phenoxybenzene, 97%, Thermo Scientific™

Product Code. 10768645
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10768645 5 g 5g
10507273 250 mg 250mg
10021962 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10768645 Supplier Thermo Scientific Maybridge Supplier No. MO07329DE
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 51632-16-7
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
InChI Key UJUNUASMYSTBSK-UHFFFAOYSA-N
Synonym 1-bromomethyl-3-phenoxybenzene, m-phenoxybenzyl bromide, 3-phenoxybenzyl bromide, benzene, 1-bromomethyl-3-phenoxy, alpha-bromo-3-phenoxytoluene, 3-phenoxybenzylbromide, unii-gd31x56z15, 3-phenyloxybenzylbromide, m-bromomethyl phenyl phenyl ether, m-phenoxybenzylbromide
PubChem CID 94544
IUPAC Name 1-(bromomethyl)-3-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=CC(=C2)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 263.13
Percent Purity 97%
Chemical Name or Material 1-(Bromomethyl)-3-phenoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.