missing translation for 'onlineSavingsMsg'
Learn More

1-Bromopyrene, 95%

Product Code. 11373437
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373437 1 g 1g
11363437 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373437 Supplier Thermo Scientific Alfa Aesar Supplier No. 018280.03
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1714-29-0 | C16H9Br | 281.152 g/mol

1-Bromopyrene is used in the cleavage of DNA in studies. It is also used in the preparation of DNA base analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1714-29-0
Molecular Formula C16H9Br
Molecular Weight (g/mol) 281.152
MDL Number MFCD00015767
InChI Key HYGLETVERPVXOS-UHFFFAOYSA-N
Synonym 1-bromo-pyrene, pyrene, bromo, pyrene, 1-bromo, bromopyrene, 1-brompyren, 1-brpy, 3-bromopyrene, 1-bromanylpyrene, pubchem14567, 1-bromopyrene
PubChem CID 159627
IUPAC Name 1-bromopyrene
SMILES C1=CC2=C3C(=C1)C=CC4=C(C=CC(=C43)C=C2)Br

Specifications

Melting Point 94°C to 95°C
Quantity 1 g
Formula Weight 281.13
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material 1-Bromopyrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.