missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Butanesulfonic acid, sodium salt, 99+%, HPLC grade
Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ion pair chromatography
Specifications
Specifications
| Absorbance | (5.5% aq. soln., pH 3) (1 cm cell), (5.5% aq. soln., pH 3) (1 cm cell) at 0nm, 0.04 max. at 250nm, 0.04 max. at 280nm, 0.1 max. at 220nm, 1 max. at 210nm |
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Melting Point | >300.0°C |
| Color | White |
| Loss on Drying | 1% max. (1 g, 105°C, 2 hrs) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99+% |
| Packaging | Glass bottle |
| Linear Formula | CH3(CH2)3SO3Na |
| Show More |
Safety and Handling
Product Identifier
- 1-Butanesulfonic acid, sodium salt
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu