missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-1-desmethylpiperazinyl-bosutinib, TRC

Product Code. 30362802
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362802 100 mg 100mg
30367052 1 g -
30366259 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362802 Supplier Toronto Research Chemicals Supplier No. TRCC423040100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380844-49-5
Molecular Formula C21H18Cl3N3O3
Molecular Weight (g/mol) 466.74
Synonym 7-(3-Chloropropoxy)-4-((2,4-dichloro-5-methoxyphenyl)amino)-6-methoxyquinoline-3-carbonitrile
IUPAC Name 7-(3-chloropropoxy)-4-(2,4-dichloro-5-methoxyanilino)-6-methoxyquinoline-3-carbonitrile
SMILES ClCCCOC(C(OC)=C1)=CC2=C1C(NC3=CC(OC)=C(Cl)C=C3Cl)=C(C#N)C=N2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C21H18Cl3N3O3/c1-28-18-9-17(14(23)7-15(18)24)27-21-12(10-25)11-26-16-8-20(30-5-3-4-22)19(29-2)6-13(16)21/h6-9,11H,3-5H2,1-2H3,(H,26,27)
Chemical Name or Material 1-Chloro-1-desmethylpiperazinyl-bosutinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.