missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,4-difluorobenzene, 98%

Product Code. p-7022627
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11452877

Brand: Thermo Scientific Alfa Aesar A15495.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1435-44-5 | C6H3ClF2 | 148.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1435-44-5
Molecular Formula C6H3ClF2
Molecular Weight (g/mol) 148.537
MDL Number MFCD00042569
InChI Key AJCSNHQKXUSMMY-UHFFFAOYSA-N
PubChem CID 137001
IUPAC Name 1-chloro-2,4-difluorobenzene
SMILES C1=CC(=C(C=C1F)F)Cl

Specifications

Melting Point -26°C
Density 1.353
Boiling Point 126°C to 127°C
Flash Point 32°C (89°F)
Odor Aromatic
Refractive Index 1.475
Quantity 10 g
UN Number UN1993
Beilstein 2081077
Synonym 2,4-difluorochlorobenzene, 1-chloro-2,4-difluoro-benzene, benzene,1-chloro-2,4-difluoro, benzene, 1-chloro-2,4-difluoro, pubchem3415, 2,4-difluorchlorbenzol, 2,4-diflurochlorobenzene, acmc-1c2st, intermediates-zcf02093, 2,4-di-fluorochlorobenzene
Formula Weight 148.54
Percent Purity 98%
Chemical Name or Material 1-Chloro-2,4-difluorobenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.