missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-iodobenzene, 99%

Product Code. 11453797
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 11453797

Brand: Thermo Scientific Alfa Aesar A18255.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 615-41-8 | C6H4ClI | 238.45 g/mol

1-Chloro-2-iodobenzene is used as a precursor and involved in the preparation of 2,2'-dichloro-biphenyl. It is also used to prepare N,N-diaryl-o-phenylenediamines and 4(5)-(2-chlorophenyl)-1H-imidazole. It reacts with lanthanum metal in the presence of trimethylchlorosilane to prepare 1-chloro-2-trimethylsilylbenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-2-iodobenzene is used as a precursor and involved in the preparation of 2,2′-dichloro-biphenyl. It is also used to prepare N,N-diaryl-o-phenylenediamines and 4(5)-(2-chlorophenyl)-1H-imidazole. It reacts with lanthanum metal in the presence of trimethylchlorosilane to prepare 1-chloro-2-trimethylsilylbenzene.

Solubility
Miscible with most common organic solvents. Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place. Keep the container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-41-8
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001033
InChI Key MPEOPBCQHNWNFB-UHFFFAOYSA-N
Synonym 2-chloroiodobenzene, o-chloroiodobenzene, o-iodochlorobenzene, 2-iodochlorobenzene, benzene, 1-chloro-2-iodo, 2-chlorophenyl iodide, 2-chloro-1-iodobenzene, 1-chloro-2-iodo-benzene, chlorjodbenzol, iodochlorobenzene
PubChem CID 11993
IUPAC Name 1-chloro-2-iodobenzene
SMILES ClC1=CC=CC=C1I

Specifications

Melting Point 1°C
Density 1.954
Boiling Point 108°C to 110°C (15 mmHg)
Flash Point 111°C (231°F)
Refractive Index 1.6345
Quantity 25 g
Beilstein 1905116
Sensitivity Light sensitive
Solubility Information Miscible with most common organic solvents. Immiscible with water.
Formula Weight 238.46
Percent Purity 99%
Chemical Name or Material 1-Chloro-2-iodobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Chloro-2-iodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Chloro-2-iodobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.