missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-fluorobenzene, 99%

Product Code. 11454474
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11454474

Brand: Thermo Scientific Alfa Aesar A13123.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 625-98-9 | C6H4ClF | 130.55 g/mol

1-Chloro-3-fluorobenzene used as medicine and liquid crystal intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-fluorobenzene used as medicine and liquid crystal intermediates.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-98-9
Molecular Formula C6H4ClF
Molecular Weight (g/mol) 130.55
MDL Number MFCD00000569
InChI Key VZHJIJZEOCBKRA-UHFFFAOYSA-N
Synonym m-chlorofluorobenzene, 3-chlorofluorobenzene, benzene, 1-chloro-3-fluoro, m-fluorochlorobenzene, 3-fluorochlorobenzene, 1-fluoro-3-chlorobenzene, 1-chloro-3-fluoro-benzene, m-fluoro chlorobenzene, pubchem3419, 3-fluoro-chlorobenzene
PubChem CID 223087
IUPAC Name 1-chloro-3-fluorobenzene
SMILES FC1=CC=CC(Cl)=C1

Specifications

Density 1.224
Boiling Point 126°C to 128°C
Flash Point 20°C (68°F)
Refractive Index 1.494
Quantity 500 g
UN Number UN1993
Beilstein 2039303
Solubility Information Not miscible in water.
Formula Weight 130.55
Percent Purity 99%
Chemical Name or Material 1-Chloro-3-fluorobenzene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.