missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-Fluorobenzene, SPEX CertiPrep™

Product Code. 16399727 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 16399727

Brand: SPEX Certiprep S944

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1-Chloro-3-fluorobenzene625-98-91000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL Amber Prescored Glass Ampule
CAS 625-98-9, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C6H4ClF
MDL Number MFCD00000569
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key VZHJIJZEOCBKRA-UHFFFAOYSA-N
SMILES FC1=CC=CC(Cl)=C1
IUPAC Name 1-chloro-3-fluorobenzene
Solvent or Matrix Methanol-P and T
Molecular Weight (g/mol) 130.55
PubChem CID 223087
Quantity 1 mL
Chemical Name or Material 1-Chloro-3-fluorobenzene
Show More Show Less

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.