missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-iodobenzene, 97%, Thermo Scientific™

Product Code. 10673245
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10673245 25 g 25g
10351615 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10673245 Supplier Thermo Scientific Maybridge Supplier No. TL00516EB
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 625-99-0
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001046
InChI Key JMLWXCJXOYDXRN-UHFFFAOYSA-N
Synonym 3-chloroiodobenzene, m-chloroiodobenzene, benzene, 1-chloro-3-iodo, 3-iodochlorobenzene, 3-chloro-1-iodobenzene, 1-chloro-3-iodo-benzene, m-chlorophenyl iodide, m-iodochlorobenzene, metachloroiodobenzene, 3-chloro-iodobenzene
PubChem CID 12269
IUPAC Name 1-chloro-3-iodobenzene
SMILES ClC1=CC=CC(I)=C1

Specifications

Density 1.9200g/mL
Assay Percent Range 97%
Linear Formula ClC6H4I
Quantity 25 g
Formula Weight 238.45
Percent Purity 97%
Chemical Name or Material 1-Chloro-3-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.