missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-iodobenzene, 98%

Product Code. 11493027
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493027 100 g 100g
11473027 5 g 5g
11483027 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493027 Supplier Thermo Scientific Alfa Aesar Supplier No. A15567.22
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 625-99-0 | C6H4ClI | 238.45 g/mol

1-Chloro-3-iodobenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-iodobenzene is used as a pharmaceutical intermediate.

Solubility
Soluble in water 67.2 mg/L.

Notes
Light sensitive. Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-99-0
Molecular Formula C6H4ClI
Molecular Weight (g/mol) 238.45
MDL Number MFCD00001046
InChI Key JMLWXCJXOYDXRN-UHFFFAOYSA-N
Synonym 3-chloroiodobenzene, m-chloroiodobenzene, benzene, 1-chloro-3-iodo, 3-iodochlorobenzene, 3-chloro-1-iodobenzene, 1-chloro-3-iodo-benzene, m-chlorophenyl iodide, m-iodochlorobenzene, metachloroiodobenzene, 3-chloro-iodobenzene
PubChem CID 12269
IUPAC Name 1-chloro-3-iodobenzene
SMILES ClC1=CC=CC(I)=C1

Specifications

Density 1.927
Boiling Point 228°C
Flash Point 102°C (216°F)
Refractive Index 1.631
Quantity 100 g
Beilstein 1904539
Sensitivity Light sensitive
Solubility Information Soluble in water 67.2mg/L.
Formula Weight 238.46
Percent Purity 98%
Chemical Name or Material 1-Chloro-3-iodobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.