missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-fluorobenzene, 98%

Product Code. 15700839 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15700839 25 mL Glass bottle 25mL
10703783 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15700839 Supplier Thermo Scientific Acros Supplier No. 208200250
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 352-33-0 | C6H4ClF | 130.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-33-0
Molecular Formula C6H4ClF
Molecular Weight (g/mol) 130.55
MDL Number MFCD00000603
InChI Key RJCGZNCCVKIBHO-UHFFFAOYSA-N
Synonym p-chlorofluorobenzene, p-fluorochlorobenzene, 4-chlorofluorobenzene, benzene, 1-chloro-4-fluoro, 1-fluoro-4-chlorobenzene, 1-chloro-4-fluoro-benzene, 4-fluorochlorobenzene, 1,4-fluorochlorobenzene, p-chloro-fluorobenzene, pubchem3420
PubChem CID 9604
IUPAC Name 1-chloro-4-fluorobenzene
SMILES FC1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -21.5°C
Color Colorless to Yellow
Density 1.2260g/mL
Boiling Point 129°C to 130°C
Flash Point 29°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4F
Refractive Index 1.4940 to 1.496
Quantity 25 mL
Beilstein 05, 201
Specific Gravity 1.226
Formula Weight 130.55
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Chloro-4-fluorobenzene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.