missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-4-fluorobenzene, 98%

Product Code. 11464664
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464664 500 g 500g
11444664 25 g 25g
11454664 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464664 Supplier Thermo Scientific Alfa Aesar Supplier No. A13690.36
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 352-33-0 | C6H4ClF | 130.55 g/mol

1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4'-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 '-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-4-fluorobenzeneis used as a primary and secondary intermediates, organic synthesis. And it is also used to prepare (4′-fluoro-biphenyl-4-yl)-methyl ether by using reagents NaH, Ni (OAc)2, 2,2 ′-bipyridyl, KI and solvents benzene, tetrahydrofuran at temperature of 63°C.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-33-0
Molecular Formula C6H4ClF
Molecular Weight (g/mol) 130.55
MDL Number MFCD00000603
InChI Key RJCGZNCCVKIBHO-UHFFFAOYSA-N
Synonym p-chlorofluorobenzene, p-fluorochlorobenzene, 4-chlorofluorobenzene, benzene, 1-chloro-4-fluoro, 1-fluoro-4-chlorobenzene, 1-chloro-4-fluoro-benzene, 4-fluorochlorobenzene, 1,4-fluorochlorobenzene, p-chloro-fluorobenzene, pubchem3420
PubChem CID 9604
IUPAC Name 1-chloro-4-fluorobenzene
SMILES FC1=CC=C(Cl)C=C1

Specifications

Melting Point -22°C
Density 1.228
Boiling Point 129°C to 130°C
Flash Point 29°C (84°F)
Refractive Index 1.495
Quantity 500 g
UN Number UN1993
Beilstein 1904542
Solubility Information Difficult to mix.
Formula Weight 130.55
Percent Purity 98%
Chemical Name or Material 1-Chloro-4-fluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.