missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloromethyl)cyclohexanecarbonitrile, 95%, Thermo Scientific™

Product Code. 10137392
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10137392 1 g Amber Glass Bottle 1g
10178204 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10137392 Supplier Thermo Scientific Maybridge Supplier No. RH01751DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 112905-95-0
Molecular Formula C8H12ClN
Molecular Weight (g/mol) 157.641
InChI Key BWQNUOFYAYIAIY-UHFFFAOYSA-N
Synonym 1-chloromethyl cyclohexanecarbonitrile, 1-chloromethyl cyclohexane-1-carbonitrile, cyclohexanecarbonitrile,1-chloromethyl, acmc-20ds23, 1-chloromethyl-cyclohexanecarbonitrile
PubChem CID 2794920
IUPAC Name 1-(chloromethyl)cyclohexane-1-carbonitrile
SMILES C1CCC(CC1)(CCl)C#N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 157.64
Percent Purity 95%
Chemical Name or Material 1-(chloromethyl)cyclohexanecarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.