missing translation for 'onlineSavingsMsg'
Learn More

1-des(piperidin-3R-amino)-1-chloro-Alogliptin, TRC

Product Code. 30350412
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350412 10 mg -
30353549 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30350412 Supplier Toronto Research Chemicals Supplier No. TRCC36547010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865758-96-9
Molecular Formula C13H10ClN3O2
Molecular Weight (g/mol) 275.69
Synonym 2-[(6-Chloro-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl)methyl]-benzonitrile, Alogliptin 6-Chloro Impurity
IUPAC Name 2-[(6-chloro-3-methyl-2,4-dioxopyrimidin-1-yl)methyl]benzonitrile
SMILES CN1C(=O)C=C(Cl)N(Cc2ccccc2C#N)C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H10ClN3O2/c1-16-12(18)6-11(14)17(13(16)19)8-10-5-3-2-4-9(10)7-15/h2-6H,8H2,1H3
Formula Weight 275.0462
Chemical Name or Material 2-[(6-Chloro-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl)methyl]-benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.