missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosanol, 98%

Product Code. 10509073 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10509073 25 g Glass bottle 25g
10072700 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10509073 Supplier Thermo Scientific Acros Supplier No. 215460250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 629-96-9 | C20H42O | 298.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-96-9
Molecular Formula C20H42O
Molecular Weight (g/mol) 298.56
MDL Number MFCD00002938
InChI Key BTFJIXJJCSYFAL-UHFFFAOYSA-N
Synonym 1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
PubChem CID 12404
ChEBI CHEBI:75627
IUPAC Name icosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 64°C to 66°C
Color White
Flash Point 194°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)19OH
Quantity 25 g
Beilstein 01, 431
Solubility Information Solubility in water: slightly soluble
Formula Weight 298.54
Percent Purity 98%
Physical Form Pellets, Powder or Chunks
Chemical Name or Material 1-Eicosanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.