missing translation for 'onlineSavingsMsg'
Learn More

1-Eth-1-ynyl-4-propylbenzene, 97%, Thermo Scientific™

Product Code. 10491854
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10491854 25 g 25g
10466642 1 g 1g
10111202 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10491854 Supplier Thermo Scientific Maybridge Supplier No. BTB09905EB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 62452-73-7
Molecular Formula C11H12
Molecular Weight (g/mol) 144.217
MDL Number MFCD00173886
InChI Key UVFFOABHOIMLNB-UHFFFAOYSA-N
Synonym 4-propylphenylacetylene, 1-eth-1-ynyl-4-propylbenzene, 4-n-propylphenylacetylene, 1-ethynyl-4-propyl-benzene, benzene, 1-ethynyl-4-propyl, pubchem10973, 4-n-propylphenyl ethine, 4-propyl phenylacetylene, 4-ethynyl-1-propylbenzene, 4-propyl-1-ethynylbenzene
PubChem CID 2775133
IUPAC Name 1-ethynyl-4-propylbenzene
SMILES CCCC1=CC=C(C=C1)C#C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HCidentical toC6H4(CH2)2CH3
Quantity 25 g
Formula Weight 144.22
Percent Purity 97%
Chemical Name or Material 1-Eth-1-ynyl-4-propylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.