missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2,3-dioxopiperazine, TRC

Product Code. 30133327
Change view
Click to view available options
Quantity:
100 mg
1 g
25 g
Unit Size:
100mg
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133327 25 g 25g
30133070 1 g 1g
17901265 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133327 Supplier Toronto Research Chemicals Supplier No. TRCE91736025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59702-31-7
Formula Weight 142.0742
InChI Formula InChI=1S/C6H10N2O2/c1-2-8-4-3-7-5(9)6(8)10/h2-4H2,1H3,(H,7,9)
IUPAC Name 1-ethylpiperazine-2,3-dione
Molecular Formula C6 H10 N2 O2
Molecular Weight (g/mol) 142.16
Percent Purity >95%
SMILES CCN1CCNC(=O)C1=O
Synonym 1-Ethylpiperazine-2,3-dione, Piperacillin Sodium Imp. E (EP)
Chemical Name or Material 1-Ethylpiperazine-2,3-dione
Grade HPLC
Show More Show Less

Specifications

Quantity 25 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.