accessibility menu, dialog, popup

UserName

CAS RN 59702-31-7

H3CNNHOO

CAS RN 59702-31-7

IUPAC Name: 1-ethylpiperazine-2,3-dione
Synonyms: 1-ethylpiperazine-2,3-dione
Molecular Weight (g/mol): 142.16
Molecular Formula: C6H10N2O2
InChi Key: ZBEKOEYCWKIMGU-UHFFFAOYSA-N
SMILES: CCN1CCNC(=O)C1=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results