missing translation for 'onlineSavingsMsg'
Learn More

1-Ethylpiperazine-2,3-dione, 97%

Product Code. 11483777
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483777 10 g 10g
11493777 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483777 Supplier Thermo Scientific Alfa Aesar Supplier No. A18248.09
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 59702-31-7 | C6H10N2O2 | 142.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59702-31-7
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.158
MDL Number MFCD00051825
InChI Key ZBEKOEYCWKIMGU-UHFFFAOYSA-N
Synonym 1-ethyl-2,3-dioxopiperazine, n-ethyl-2,3-dioxopiperazine, 1-ethyl-2,3-piperazinedione, 2,3-piperazinedione, 1-ethyl, 4-ethyl-2,3-dioxopiperazine, 4-ethyl-2,3-piperazinedione, ccris 8931, 4-ethyl-2,3-dioxo-piperazine, 4-ethyl-2,3-dioxypiperazine, n-ethylpiperazine-2,3-dione
PubChem CID 108812
IUPAC Name 1-ethylpiperazine-2,3-dione
SMILES CCN1CCNC(=O)C1=O

Specifications

Melting Point 113°C to 124°C
Quantity 10 g
Beilstein 879076
Formula Weight 142.16
Percent Purity 97%
Chemical Name or Material 1-Ethylpiperazine-2,3-dione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.