missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-2-iodobenzene, 99%, stab. with copper

Product Code. 11439593
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439593 500 g 500g
11419593 25 g 25g
11429593 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11439593 Supplier Thermo Scientific Alfa Aesar Supplier No. A12411.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 348-52-7 | C6H4FI | 222.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-52-7
Molecular Formula C6H4FI
Molecular Weight (g/mol) 222.00
MDL Number MFCD00001031
InChI Key TYHUGKGZNOULKD-UHFFFAOYSA-N
Synonym 2-fluoroiodobenzene, o-fluoroiodobenzene, o-iodofluorobenzene, benzene, 1-fluoro-2-iodo, 2-iodofluorobenzene, 3-fluoro-2-iodobenzene, 1-fluoro-2-iodo-benzene, 2-fluoro-iodobenzene, 2-fluoro-1-iodobenzene
PubChem CID 67673
IUPAC Name 1-fluoro-2-iodobenzene
SMILES FC1=CC=CC=C1I

Specifications

Melting Point -43°C to -40°C
Density 1.903
Boiling Point 188°C to 189°C
Flash Point 71°C (159°F)
Refractive Index 1.59
Quantity 500 g
Beilstein 2039770
Sensitivity Light sensitive
Formula Weight 222
Percent Purity 99%
Chemical Name or Material 1-Fluoro-2-iodobenzene, Stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.