missing translation for 'onlineSavingsMsg'
Learn More

1-(Fmoc-amino)cyclohexanecarboxylic acid, 98+%

Product Code. 11389785
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389785 5 g 5g
11379785 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389785 Supplier Thermo Scientific Alfa Aesar Supplier No. L19810.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 162648-54-6 | C22H23NO4 | 365.43 g/mol

1-(Fmoc-amino)cyclohexanecarboxylic acid is an API so it is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals industry. Also used as raw materials for sodium fluosilicate, potassium fluosilicate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Fmoc-amino)cyclohexanecarboxylic acid is an API so it is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals industry. Also used as raw materials for sodium fluosilicate, potassium fluosilicate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162648-54-6
Molecular Formula C22H23NO4
Molecular Weight (g/mol) 365.43
MDL Number MFCD00273464
InChI Key VCIVAWBKUQJNSX-UHFFFAOYSA-N
Synonym 1-fmoc-amino cyclohexanecarboxylic acid, fmoc-ac6c-oh, fmoc-homocycloleucine, 1-9h-fluoren-9-yl methoxy carbonyl amino cyclohexanecarboxylic acid, fmoc-1-aminocyclohexanecarboxylic acid, 1-9h-fluoren-9-ylmethoxy carbonyl amino cyclohexane-1-carboxylic acid, acmc-209dor, fmoc-nh 1 chex-oh, 1-9h-fluoren-9-ylmethoxycarbonylamino cyclohexane-1-carboxylic acid
PubChem CID 2756093
IUPAC Name 1-(9H-fluoren-9-ylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
SMILES OC(=O)C1(CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 185°C to 187°C (decomposition)
Quantity 5 g
Beilstein 7887646
Solubility Information Insoluble in water.
Formula Weight 365.43
Percent Purity ≥98%
Chemical Name or Material 1-(Fmoc-amino)cyclohexanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.