missing translation for 'onlineSavingsMsg'
Learn More

(1-Hexyl)triphenylphosphonium bromide, 98%

Product Code. 11484914
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11484914

Brand: Thermo Scientific Alfa Aesar A13826.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4762-26-9 | C24H28BrP | 427.37 g/mol

(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Hexyl)triphenylphosphonium bromide can be used as catalyst in pharmaceutical research. It is used in organic synthesis. Reactive wittg reagent used in prostaglandin synthesis.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Store under inert gas. Protect from light. Store away from oxidizing materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4762-26-9
Molecular Formula C24H28BrP
Molecular Weight (g/mol) 427.37
MDL Number MFCD00011860
InChI Key PWDFZWZPWFYFTC-UHFFFAOYSA-M
Synonym hexyltriphenylphosphonium bromide, n-hexyl-triphenylphosphonium bromide, phosphonium, hexyltriphenyl-, bromide, hexyltriphenylphosphoniumbromide, hexyltriphenylphosphanium bromide, n-hexyl triphenylphosphonium bromide, c24h28p.br, acmc-209k9x, n-hexyltriphenylphosphonium bromide, hexyl triphenyl phosphanium bromide
PubChem CID 2724569
SMILES [Br-].CCCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 199°C to 202°C
Quantity 500 g
Beilstein 3580731
Sensitivity Hygroscopic
Solubility Information Soluble in water and methanol.
IUPAC Name hexyl(triphenyl)phosphanium;bromide
Formula Weight 427.37
Percent Purity 98%
Chemical Name or Material (1-Hexyl)triphenylphosphonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.