missing translation for 'onlineSavingsMsg'
Learn More

1-Hexylboronic acid pinacol ester, 98%

Product Code. 11367409
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367409 1 g 1g
11377409 5 g 5g
11387409 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11367409 Supplier Thermo Scientific Alfa Aesar Supplier No. H27831.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 86308-26-1 | C12H25BO2 | 212.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86308-26-1
Molecular Formula C12H25BO2
Molecular Weight (g/mol) 212.14
MDL Number MFCD08457665
InChI Key LDPMCSWZEGKWLC-UHFFFAOYSA-N
Synonym hexylboronic acid pinacol ester, 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborole, 1,3,2-dioxaborolane, 2-hexyl-4,4,5,5-tetramethyl
PubChem CID 10921795
IUPAC Name 2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)CCCCCC

Specifications

Density 1.167
Flash Point 84°C (183°F)
Refractive Index 1.426
Quantity 1 g
Formula Weight 212.14
Percent Purity 98%
Chemical Name or Material 1-Hexylboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.