missing translation for 'onlineSavingsMsg'
Learn More

1-Indanol, 98%

Product Code. 10656202 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10656202 10 g Glass bottle 10g
10337940 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10656202 Supplier Thermo Scientific Acros Supplier No. 186110100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6351-10-6 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10-6-6351
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003797
InChI Key YIAPLDFPUUJILH-UHFFFAOYNA-N
Synonym 1-indanol, indanol, indan-1-ol, 1-hydroxyhydrindene, 1h-inden-1-ol, 2,3-dihydro, 1h-indenol, 2,3-dihydro, 1-hydroxyindan, 1-hydroxyindane, 1-indanole, --indanol
PubChem CID 22819
ChEBI CHEBI:16697
IUPAC Name 2,3-dihydro-1H-inden-1-ol
SMILES C1CC2=CC=CC=C2C1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 50°C to 54°C
Color White to Yellow
Boiling Point 260°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 06, 574
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in ethanol,benzene and chloroform,slightly soluble in petroleum ether
Formula Weight 134.18
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 1-Indanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Indanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.