missing translation for 'onlineSavingsMsg'
Learn More

1-Indanol, 98%

Product Code. 11420378
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420378 25 g 25g
11410378 5 g 5g
11424170 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420378 Supplier Thermo Scientific Alfa Aesar Supplier No. B24536.14
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6351-10-6 | C9H10O | 134.18 g/mol

1-Indanol derivative was found to be an efficient ligand for the palladium-catalyzed asymmetric Heck reaction. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Indanol derivative was found to be an efficient ligand for the palladium-catalyzed asymmetric Heck reaction. It is used as pharmaceutical intermediate.

Solubility
Soluble in water 10 g/L at 20°C, ethanol, benzene.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10-6-6351
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003797
InChI Key YIAPLDFPUUJILH-UHFFFAOYNA-N
Synonym 1-indanol, indanol, indan-1-ol, 1-hydroxyhydrindene, 1h-inden-1-ol, 2,3-dihydro, 1h-indenol, 2,3-dihydro, 1-hydroxyindan, 1-hydroxyindane, 1-indanole, --indanol
PubChem CID 22819
ChEBI CHEBI:16697
IUPAC Name 2,3-dihydro-1H-inden-1-ol
SMILES C1CC2=CC=CC=C2C1O

Specifications

Melting Point 50°C to 54°C
Boiling Point 128°C (12 mmHg)
Flash Point 145°C (293°F)
Quantity 25 g
Beilstein 2042960
Solubility Information Soluble in water 10g/L at 20°C,ethanol,benzene.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 1-Indanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.