missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Iodo-2,4-dinitrobenzene, 98%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.
Solubility
Insoluble in water.
Notes
Light sensitive. Store away from oxidizing agents, bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under dark.
It is employed in the cloning and characterization of a biotic-stress-inducible glutathione transferase from Phaseolus vulgaris.
Solubility
Insoluble in water.
Notes
Light sensitive. Store away from oxidizing agents, bases and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under dark.
Specifications
Specifications
| Melting Point | 87°C to 90°C |
| Quantity | 25 g |
| Beilstein | 1113584 |
| Sensitivity | Light sensitive |
| Solubility Information | Insoluble in water. |
| Formula Weight | 294.01 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-Iodo-2,4-dinitrobenzene |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction