missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-2-nitrobenzene, 97%, Thermo Scientific Chemicals

Product Code. 10606942 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10606942 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10606942 Supplier Thermo Scientific Acros Supplier No. 159450250
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 609-73-4 | Molecular Formula: C6H4INO2 | Molecular Weight: 249.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-73-4
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.01
MDL Number MFCD00007088
InChI Key JXMZUNPWVXQADG-UHFFFAOYSA-N
Synonym 2-iodonitrobenzene, o-iodonitrobenzene, o-nitroiodobenzene, benzene, 1-iodo-2-nitro, 2-nitroiodobenzene, 2-iodo-1-nitrobenzene, 1-iodo-2-nitro-benzene, 2-iodo-nitrobenzene, 2-nitro-iodobenzene, benzene, iodonitro
PubChem CID 69115
IUPAC Name 1-iodo-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1I

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 47°C to 52°C
Color Brown-Yellow to Green-Yellow
Boiling Point 288°C to 289°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4NO2
Quantity 25 g
Beilstein 05, 252
Solubility Information Solubility in water: insoluble
Formula Weight 249.01
Percent Purity 97%
Physical Form Granular Powder
Chemical Name or Material 1-Iodo-2-nitrobenzene, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Iodo-2-nitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.