missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-isopropylbenzene, 97%

Product Code. 11427156
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427156 5 g 5g
11437156 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427156 Supplier Thermo Scientific Alfa Aesar Supplier No. B25476.06
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 17356-09-1 | C9H11I | 246.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17356-09-1
Molecular Formula C9H11I
Molecular Weight (g/mol) 246.091
MDL Number MFCD00013711
InChI Key PQJOSEVTIKYWLH-UHFFFAOYSA-N
Synonym 1-iodo-4-isopropylbenzene, p-iodocumene, 4-iodoisopropylbenzene, 2-4'-iodophenyl propane, 4-iodocumene, p-iodoisopropylbenzene, 1-iodo-4-propan-2-yl benzene, 4-isopropyliodobenzene, benzene,1-iodo-4-1-methylethyl, benzene, 1-iodo-4-1-methylethyl
PubChem CID 87073
IUPAC Name 1-iodo-4-propan-2-ylbenzene
SMILES CC(C)C1=CC=C(C=C1)I

Specifications

Density 1.525
Boiling Point 236°C to 238°C
Refractive Index 1.578
Quantity 5 g
Beilstein 2205506
Sensitivity Light sensitive
Formula Weight 246.09
Percent Purity 97%
Chemical Name or Material 1-Iodo-4-isopropylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.