missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-nitrobenzene, 98+%

Product Code. 11426826
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11426826 100 g 100g
11406826 5 g 5g
11416826 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426826 Supplier Thermo Scientific Alfa Aesar Supplier No. B23950.22
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 636-98-6 | C6H4INO2 | 249.007 g/mol

1-Iodo-4-nitrobenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Iodo-4-nitrobenzene is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-98-6
Molecular Formula C6H4INO2
Molecular Weight (g/mol) 249.007
MDL Number MFCD00007299
InChI Key SCCCFNJTCDSLCY-UHFFFAOYSA-N
Synonym benzene, 1-iodo-4-nitro, p-iodonitrobenzene, 4-iodonitrobenzene, p-nitroiodobenzene, 4-nitroiodobenzene, 4-iodo-nitrobenzene, p-nitrophenyl iodide, 1-iodo-4-nitro-benzene, 4-iodo-1-nitrobenzene, 4-nitro-1-iodobenzene
PubChem CID 12495
IUPAC Name 1-iodo-4-nitrobenzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])I

Specifications

Melting Point 172°C to 177°C
Boiling Point 288°C to 290°C
Quantity 100 g
Beilstein 1100378
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 249.01
Percent Purity ≥98%
Chemical Name or Material 1-Iodo-4-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.