missing translation for 'onlineSavingsMsg'
Learn More

1-Iodo-4-(trifluoromethoxy)benzene, 98%

Product Code. 11445667
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445667 25 g 25g
11425667 1 g 1g
11435667 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11445667 Supplier Thermo Scientific Alfa Aesar Supplier No. B23628.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 103962-05-6 | C7H4F3IO | 288.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103962-05-6
Molecular Formula C7H4F3IO
Molecular Weight (g/mol) 288.01
MDL Number MFCD00042411
InChI Key RTUDBROGOZBBIC-UHFFFAOYSA-N
Synonym 1-iodo-4-trifluoromethoxy benzene, 4-trifluoromethoxy iodobenzene, 4-iodotrifluoromethoxybenzene, 1-iado-4-trifluoromethoxy benzene, p-trifluoromethoxy iodobenzene, 4-trifluoromethoxy-iodobenzene, 1-iodo-4-trifluoromethoxy-benzene, benzene, 1-iodo-4-trifluoromethoxy, alpha,alpha,alpha-trifluoro-4-iodoanisole, 4-iodo-alpha,alpha,alpha-trifluoromethoxybenzene
PubChem CID 2777294
IUPAC Name 1-iodo-4-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=C(I)C=C1

Specifications

Density 1.909
Boiling Point 72°C to 74°C (15 mmHg)
Flash Point 77°C (170°F)
Refractive Index 1.5033
Quantity 25 g
Beilstein 6193794
Sensitivity Light sensitive
Formula Weight 288.01
Percent Purity 98%
Chemical Name or Material 1-Iodo-4-(trifluoromethoxy)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.