missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Iodoheptane, 98%, stab. with copper
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Description
1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.
Solubility
Not miscible or difficult to mix in water.
Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
1-Iodoheptane is used in the functionalization of symmetrical star-shaped molecules with carborane clusters.
Solubility
Not miscible or difficult to mix in water.
Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. It is sensitive to light. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | -48°C |
| Density | 1.379 |
| Boiling Point | 203°C to 205°C |
| Flash Point | 78°C (174°F) |
| Refractive Index | 1.49 |
| Quantity | 50 g |
| Sensitivity | Light sensitive |
| Solubility Information | Not miscible or difficult to mix in water. |
| Formula Weight | 226.1 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
Product Identifier
- 1-Iodoheptane
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 1-Iodoheptane
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction