missing translation for 'onlineSavingsMsg'
Learn More

1-Isoquinolinecarbonitrile, 98%, Thermo Scientific Chemicals

Product Code. 13408678 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 13408678

Brand: Thermo Scientific Acros 449570050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1198-30-7
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.17
InChI Key HJHXYSBRTVFEDD-UHFFFAOYSA-N
Synonym 1-cyanoisoquinoline, 1-isoquinolinecarbonitrile, isoquinaldonitrile, isoquinolinecarbonitrile, 1-cyano-isoquinoline, isoquinaldinic nitrile, pubchem6269, maybridge1_002114, timtec-bb sbb002487
PubChem CID 306057
IUPAC Name isoquinoline-1-carbonitrile
SMILES C1=CC=C2C(=C1)C=CN=C2C#N

Specifications

Melting Point 90.0°C to 92.0°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 154.17
Percent Purity 98%
Chemical Name or Material 1-Isoquinolinecarbonitrile
compliance-icons
Product Identifier
  • 1-Isoquinolinecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.