missing translation for 'onlineSavingsMsg'
Learn More

1-(Mesitylene-2-sulfonyl)-3-nitro-1,2,4-triazole, 98%

Product Code. 10388400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10388400 5 g Glass bottle 5g
10184700 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10388400 Supplier Thermo Scientific Acros Supplier No. 221310050
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 74257-00-4 | C11H12N4O4S | 296.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74257-00-4
Molecular Formula C11H12N4O4S
Molecular Weight (g/mol) 296.30
MDL Number MFCD00009754
InChI Key SFYDWLYPIXHPML-UHFFFAOYSA-N
Synonym 1-mesitylsulfonyl-3-nitro-1h-1,2,4-triazole, 1-mesitylene-2-sulfonyl-3-nitro-1,2,4-triazole, msnt, 1-mesitylene-2-sulfonyl-3-nitro-1h-1,2,4-triazole, 3-nitro-1-2,4,6-trimethylphenyl sulfonyl-1,2,4-triazole, 1-2-mesitylenesulfonyl-3-nitro-1h-1,2,4-triazole, 2,4,6-trimethylphenyl-3-nitro-1,2,4-triazol-1-yl sulfone, 3-nitro-1-2,4,6-trimethylbenzenesulfonyl-1h-1,2,4-triazole, 1h-1,2,4-triazole, 3-nitro-1-2,4,6-trimethylphenyl sulfonyl
PubChem CID 716901
IUPAC Name 3-nitro-1-(2,4,6-trimethylphenyl)sulfonyl-1,2,4-triazole
SMILES CC1=CC(C)=C(C(C)=C1)S(=O)(=O)N1C=NC(=N1)[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 132.0°C to 134.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 296.29
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-(Mesitylene-2-sulfonyl)-3-nitro-1, 2, 4-triazole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.